Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102371.1 | NP_001095841.1 | 5 | 1197 | ||
NM_024955.5 | NP_079231.4 | 5 | 1236 | |||
XM_017028941.1 | XP_016884430.1 | 4 | 869 | |||
XM_017028942.1 | XP_016884431.1 | 4 | 966 | |||
GenBank mRNA | AK025714.1 | - | 5 | 1136 | ||
AK026975.1 | - | 2 | 317 | |||
AK225347.1 | - | 5 | 1136 | |||
AK315555.1 | - | 5 | 1139 | |||
BC015726.1 | - | 5 | 1120 | 4866761 | ||
BC027716.2 | - | 5 | 1215 | 4127473 | ||
BC040351.1 | - | 5 | 1159 | 3873448 | ||
CR456444.1 | - | 4 | 1211 |