Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NRSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323680.1 | NP_001310609.1 | 4 | 963 | ||
NM_001323681.1 | NP_001310610.1 | 4 | 954 | |||
NM_001323682.1 | NP_001310611.1 | 5 | 1020 | |||
NM_001323683.1 | NP_001310612.1 | 5 | 1014 | |||
NM_001323684.1 | NP_001310613.1 | 5 | 1042 | |||
NM_001323685.1 | NP_001310614.1 | 4 | 683 | |||
NM_024958.2 | NP_079234.1 | 4 | 941 | |||
NR_136649.1 | 3 | 746 | ||||
XM_006723630.3 | XP_006723693.1 | 4 | 1364 | |||
XM_011529360.1 | XP_011527662.1 | 4 | 647 | |||
XM_011529362.2 | XP_011527664.1 | 5 | 654 | |||
XM_011529363.2 | XP_011527665.1 | 4 | 632 | |||
XM_017028073.1 | XP_016883562.1 | 4 | 586 | |||
XM_017028074.1 | XP_016883563.1 | 4 | 640 | |||
XM_017028075.1 | XP_016883564.1 | 4 | 812 | |||
GenBank mRNA | AK026982.1 | - | 4 | 554 | ||
AK290527.1 | - | 4 | 918 | |||
AL136915.1 | - | 3 | 536 | |||
BC001963.1 | - | 3 | 521 | 3454427 | ||
BE792891.1 | - | 2 | 305 | |||
CD102518.1 | - | 4 | 572 | |||
CR533441.1 | - | 2 | 403 |
Gene Symbol: LOC101929937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028182.1 | XP_016883671.1 | 1 | 112 |