Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025092.4 | NP_079368.3 | 4, 5 | 1029 | ||
XM_011520383.2 | XP_011518685.1 | 7, 8 | 1955 | |||
XM_011520384.2 | XP_011518686.1 | 6, 7 | 2066 | |||
XM_011520385.2 | XP_011518687.1 | 6, 7 | 2069 | |||
XM_011520386.2 | XP_011518688.1 | 6, 7 | 2069 | |||
XM_011520387.2 | XP_011518689.1 | 6, 7 | 2069 | |||
XM_017018355.1 | XP_016873844.1 | 5, 6 | 1407 | |||
GenBank mRNA | AK026288.1 | - | 1, 2 | 440 | ||
AK074155.1 | - | 4 | 1894 | |||
AK090428.1 | - | 1 | 425 | |||
BC066641.1 | - | 4, 5 | 949 | |||
BC109257.2 | - | 1, 2 | 428 | 40024867 | ||
BC109258.2 | - | 1, 2 | 428 | 40024874 | ||
BP262327.1 | - | 1, 2 | 439 |