Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124756.2 | NP_001118228.1 | 3, 4 | 590 | ||
NR_134983.1 | 3, 4 | 590 | ||||
GenBank mRNA | DR001377.1 | - | 3, 4 | 578 | ||
EU190483.1 | - | 3, 4 | 526 |