Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 3 | 1149 | ||
NM_001317952.1 | NP_001304881.1 | 2 | 707 | |||
NM_030767.5 | NP_110394.3 | 3 | 1094 | |||
XM_005252244.2 | XP_005252301.1 | 5 | 1266 | |||
XM_005252245.1 | XP_005252302.1 | 4 | 1124 | |||
XM_005252247.4 | XP_005252304.1 | 3 | 1145 | |||
XM_005252248.2 | XP_005252305.1 | 3 | 1262 | |||
XM_006717294.1 | XP_006717357.1 | 3 | 1078 | |||
XM_006717295.2 | XP_006717358.1 | 3 | 1262 | |||
XM_011519063.2 | XP_011517365.1 | 2 | 757 | |||
XM_011519064.2 | XP_011517366.1 | 3 | 1262 | |||
XM_011519065.2 | XP_011517367.2 | 3 | 737 | |||
XM_011519066.2 | XP_011517368.1 | 3 | 1262 | |||
XM_017015172.1 | XP_016870661.1 | 4 | 864 | |||
GenBank mRNA | AB075848.1 | - | 3 | 1267 | ||
AK024431.1 | - | 1 | 42 | |||
AK074040.1 | - | 1 | 90 | |||
AK126136.1 | - | 3 | 1078 | |||
AK131082.2 | - | 3 | 1149 | |||
AK160382.1 | - | 3 | 1149 | |||
AY703039.1 | - | 4 | 749 | |||
AY703040.1 | - | 2 | 707 | |||
AY703041.1 | - | 4 | 463 | |||
AY703042.1 | - | 2 | 276 | |||
AY703043.1 | - | 2 | 1067 | |||
AY703044.1 | - | 1 | 761 | |||
BC055285.1 | - | 3 | 1094 | 6143155 |