Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178044.1 | NP_001171515.1 | 15 | 1538 | ||
NM_001178045.1 | NP_001171516.1 | 16 | 1667 | |||
NM_025257.2 | NP_079533.2 | 16 | 1664 | |||
GenBank mRNA | AK027397.1 | - | 16 | 1615 | ||
AK222998.1 | - | 16 | 1615 | |||
AK300550.1 | - | 16 | 1667 | |||
AK301596.1 | - | 15 | 1489 | |||
AK314673.1 | - | 16 | 1619 | |||
AL833009.1 | - | 12 | 1357 | |||
AY358457.1 | - | 4 | 431 | |||
BC014659.1 | - | 16 | 1603 | 4644272 | ||
KU175229.1 | - | 16 | 1598 |