Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098536.1 | NP_001092006.1 | 18 | 2326 | ||
NM_003481.2 | NP_003472.2 | 18 | 2251 | |||
GenBank mRNA | BC004889.2 | - | 18 | 2230 | 3936781 | |
BC005139.2 | - | 18 | 2297 | 3506801 | ||
U35116.1 | - | 18 | 2241 | |||
U47927.1 | - | 18 | 2319 | |||
X91349.1 | - | 18 | 2296 |