Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030640.2 | NP_085143.1 | 7 | 2359 | ||
XM_005253488.2 | XP_005253545.1 | 6 | 2520 | |||
XM_006719155.2 | XP_006719218.1 | 7 | 2087 | |||
XM_011520856.1 | XP_011519158.1 | 8 | 2163 | |||
XM_011520857.1 | XP_011519159.1 | 8 | 2808 | |||
XM_017019988.1 | XP_016875477.1 | 6 | 1923 | |||
GenBank mRNA | AB051487.1 | - | 6 | 1779 | ||
AB052156.1 | - | 7 | 2159 | |||
AF506796.1 | - | 7 | 2228 | |||
AK055973.1 | - | 6 | 1651 | |||
AK098310.1 | - | 2 | 579 | |||
AY038927.1 | - | 5 | 1459 | |||
BC031643.1 | - | 6 | 1450 | |||
BC042101.1 | - | 6 | 1918 | 4400399 | ||
BC109234.1 | - | 7 | 2189 | 40012251 | ||
BC109235.1 | - | 7 | 2189 | 40012256 |