Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136043.2 | NP_001129515.1 | 4 | 566 | ||
NM_001207058.1 | NP_001193987.1 | 4 | 545 | |||
NM_001207059.1 | NP_001193988.1 | 3 | 425 | |||
NR_045104.1 | 4 | 709 | ||||
NR_045105.2 | 4 | 963 | ||||
NR_045106.2 | 4 | 870 | ||||
NR_045901.1 | 3 | 601 | ||||
XM_006720708.3 | XP_006720771.1 | 4 | 645 | |||
XM_006720709.3 | XP_006720772.1 | 4 | 710 | |||
XM_006720711.2 | XP_006720774.1 | 3 | 569 | |||
XM_011522101.2 | XP_011520403.1 | 4 | 391 | |||
XM_017022644.1 | XP_016878133.1 | 4 | 790 | |||
GenBank mRNA | AK027475.1 | - | 4 | 551 | ||
AK293207.1 | - | 3 | 378 | |||
AK296134.1 | - | 4 | 936 | |||
AK301033.1 | - | 4 | 408 | |||
AK301222.1 | - | 3 | 554 | |||
AK301742.1 | - | 4 | 387 | |||
AK310857.1 | - | 4 | 555 | |||
AL136662.1 | - | 4 | 791 | |||
BC007991.2 | - | 4 | 536 | 3140504 | ||
DC298728.1 | - | 3 | 378 | |||
DC384988.1 | - | 4 | 462 |