Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319303.1 | NP_001306232.1 | 3 | 1505 | ||
NM_030929.4 | NP_112191.2 | 3 | 1450 | |||
NR_135067.1 | 2 | 888 | ||||
XM_005270194.3 | XP_005270251.1 | 3 | 1551 | |||
XM_017016714.1 | XP_016872203.1 | 2 | 529 | |||
XM_017016715.1 | XP_016872204.1 | 2 | 529 | |||
GenBank mRNA | AF333487.1 | - | 2 | 615 | ||
AK172862.1 | - | 3 | 701 | |||
AK172864.1 | - | 3 | 1504 | |||
AK225949.1 | - | 3 | 702 | |||
AY014271.1 | - | 2 | 615 | |||
AY359087.1 | - | 3 | 706 | |||
BC007758.2 | - | 3 | 851 | 4122366 | ||
BI766345.1 | - | 2 | 161 | |||
CX871488.1 | - | 2 | 179 |