Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033342.3 | NP_203128.1 | 2 | 758 | ||
NM_203293.2 | NP_976038.1 | 2 | 758 | |||
NM_203294.1 | NP_976039.1 | 2 | 372 | |||
NM_203295.1 | NP_976040.1 | 2 | 372 | |||
NM_203296.1 | NP_976041.1 | 2 | 649 | |||
XM_017009903.1 | XP_016865392.1 | 2 | 729 | |||
XM_017009904.1 | XP_016865393.1 | 2 | 1465 | |||
GenBank mRNA | AF220032.1 | - | 2 | 758 | ||
AF396651.1 | - | 2 | 635 | |||
AF396652.1 | - | 2 | 372 | |||
AF396653.1 | - | 2 | 372 | |||
AF396654.1 | - | 2 | 433 | |||
AK314872.1 | - | 2 | 621 | |||
BC011567.1 | - | 2 | 611 | |||
BC080553.1 | - | 2 | 588 | |||
BC132863.1 | - | 2 | 637 | 40146885 | ||
BC132867.1 | - | 2 | 637 | 40146889 | ||
BM970007.1 | - | 1 | 478 |