Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198868.1 | NP_001185797.1 | 9 | 1142 | ||
NM_001198869.1 | NP_001185798.1 | 9 | 1111 | |||
NM_005186.3 | NP_005177.2 | 9 | 1127 | |||
NR_040008.1 | 8 | 1057 | ||||
XM_006718698.1 | XP_006718761.1 | 9 | 1042 | |||
XM_011545292.1 | XP_011543594.1 | 9 | 1053 | |||
GenBank mRNA | AK301538.1 | - | 8 | 949 | ||
AK311001.1 | - | 8 | 697 | |||
AK315556.1 | - | 9 | 1056 | |||
AY550975.1 | - | 8 | 950 | |||
BC008751.2 | - | 9 | 1078 | 3347480 | ||
BC017200.2 | - | 9 | 964 | 3629894 | ||
BC075862.1 | - | 9 | 1045 | 5223130 | ||
X04366.1 | - | 9 | 1088 |