Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004188.6 | NP_004179.3 | 9 | 1369 | ||
XM_006717297.3 | XP_006717360.1 | 5 | 860 | |||
XM_011519068.2 | XP_011517370.1 | 7 | 1260 | |||
XM_011519069.2 | XP_011517371.1 | 6 | 1030 | |||
XM_011519070.2 | XP_011517372.1 | 7 | 1260 | |||
XM_017015175.1 | XP_016870664.1 | 6 | 990 | |||
GenBank mRNA | AF081946.1 | - | 4 | 544 | ||
AY428733.1 | - | 5 | 695 | |||
BC043371.1 | - | 9 | 1369 | 5170920 | ||
BC048254.1 | - | 4 | 575 | |||
CR536546.1 | - | 4 | 544 |