Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201474.1 | NP_001188403.1 | 3 | 504 | ||
NM_001201475.1 | NP_001188404.1 | 3 | 571 | |||
NM_031446.4 | NP_113634.3 | 4 | 492 | |||
XM_005258364.4 | XP_005258421.1 | 4 | 497 | |||
GenBank mRNA | AF277182.1 | - | 4 | 351 | ||
AF277189.1 | - | 4 | 351 | |||
AW967900.1 | - | 3 | 464 | |||
BC108730.1 | - | 4 | 452 | 5187778 | ||
BC119765.1 | - | 4 | 443 | 40119355 | ||
BC119766.1 | - | 4 | 442 | 40119357 | ||
BX460890.2 | - | 4 | 818 |