Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISPLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031476.3 | NP_113664.1 | 5 | 814 | ||
XM_005256190.1 | XP_005256247.1 | 6 | 1069 | |||
GenBank mRNA | AK027395.1 | - | 5 | 778 | ||
AK308396.1 | - | 6 | 741 | |||
AL136861.1 | - | 5 | 791 | |||
AY358790.1 | - | 5 | 823 | |||
BC063012.1 | - | 5 | 792 | 5217820 |