Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098529.1 | NP_001091999.1 | 2 | 927 | ||
NM_032243.5 | NP_115619.4 | 2 | 724 | |||
XM_017026041.1 | XP_016881530.1 | 3 | 598 | |||
GenBank mRNA | AF080095.1 | - | 2 | 457 | ||
AK097656.1 | - | 2 | 677 | |||
AK303045.1 | - | 2 | 684 | |||
BC050132.1 | - | 2 | 512 | 5164861 | ||
BM554364.1 | - | 2 | 632 | |||
EF560747.1 | - | 2 | 603 |