Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303481.1 | NP_001290410.1 | 3 | 797 | ||
NM_003575.3 | NP_003566.1 | 3 | 797 | |||
XM_006716151.3 | XP_006716214.1 | 3 | 791 | |||
GenBank mRNA | AK299280.1 | - | 3 | 709 | ||
AK316282.1 | - | 3 | 684 | |||
AY927504.1 | - | 1 | 61 | |||
AY927505.1 | - | 2 | 470 | |||
BC031686.1 | - | 2 | 341 | |||
BC041704.1 | - | 3 | 524 | |||
BC055287.1 | - | 3 | 587 | |||
BC073805.1 | - | 3 | 690 | 6645861 | ||
D30612.1 | - | 3 | 810 |