Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032414.2 | NP_115790.1 | 1 | 78 | ||
GenBank mRNA | AF333024.1 | - | 1 | 81 | ||
AY029225.1 | - | 1 | 75 | |||
AY358683.1 | - | 1 | 117 | |||
BC025399.1 | - | 1 | 55 | 4826496 | ||
BM544315.1 | - | 1 | 104 |