Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM2 SIRNA Assays ›
Gene Symbol:
ABLIM2
Gene Name:
actin binding LIM protein family member 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 7965310 - 8158832 on Build GRCh38
UniGene ID:
Hs.233404
Species:
Human
siRNA ID
HSS189072
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130083.1
NP_001123555.1
15
1598
NM_001130084.1
NP_001123556.1
14
1499
NM_001130085.1
NP_001123557.1
14
1499
NM_001130086.1
NP_001123558.1
12
1343
NM_001130087.1
NP_001123559.1
13
1376
NM_001130088.1
NP_001123560.1
13
1376
NM_001286688.1
NP_001273617.1
9
860
NM_032432.4
NP_115808.3
12
1343
XM_005248014.4
XP_005248071.1
17
1722
XM_005248017.4
XP_005248074.1
17
1722
XM_005248018.4
XP_005248075.1
16
1601
XM_005248019.4
XP_005248076.1
15
1568
XM_005248020.4
XP_005248077.1
15
1568
XM_005248021.4
XP_005248078.1
16
1600
XM_005248022.4
XP_005248079.1
14
1469
XM_005248023.4
XP_005248080.1
15
1568
XM_005248024.4
XP_005248081.1
14
1468
XM_005248027.4
XP_005248084.1
12
1312
XM_005248028.4
XP_005248085.1
13
1342
XM_005248029.4
XP_005248086.1
12
1311
XM_005248030.4
XP_005248087.1
12
1311
XM_005248031.4
XP_005248088.1
13
1346
XM_006713923.3
XP_006713986.1
17
1722
XM_006713924.3
XP_006713987.1
16
1601
XM_006713925.3
XP_006713988.1
16
1602
XM_006713927.3
XP_006713990.1
13
1345
XM_011513586.2
XP_011511888.1
17
1726
XM_017008706.1
XP_016864195.1
17
1723
XM_017008707.1
XP_016864196.1
17
1732
XM_017008708.1
XP_016864197.1
16
1688
XM_017008709.1
XP_016864198.1
16
1623
XM_017008710.1
XP_016864199.1
15
1502
XM_017008711.1
XP_016864200.1
15
1589
XM_017008712.1
XP_016864201.1
15
1589
XM_017008713.1
XP_016864202.1
15
1567
XM_017008714.1
XP_016864203.1
15
1579
XM_017008715.1
XP_016864204.1
14
1514
XM_017008716.1
XP_016864205.1
15
1501
XM_017008717.1
XP_016864206.1
15
1501
XM_017008718.1
XP_016864207.1
15
1513
XM_017008719.1
XP_016864208.1
14
1468
XM_017008720.1
XP_016864209.1
13
1344
XM_017008721.1
XP_016864210.1
12
1311
XM_017008722.1
XP_016864211.1
13
1414
XM_017008723.1
XP_016864212.1
16
1612
XM_017008724.1
XP_016864213.1
13
1357
XM_017008725.1
XP_016864214.1
12
1324
XM_017008726.1
XP_016864215.1
11
1061
XM_017008727.1
XP_016864216.1
11
1061
GenBank mRNA
AB058711.1
-
13
1258
AJ748600.1
-
14
1421
AJ748601.1
-
14
1421
AK094754.1
-
13
1355
AK094798.1
-
9
830
AL834195.1
-
3
710
BC067214.1
-
13
1388
BC122567.1
-
12
1294
40085570
BM718728.1
-
2
243
DQ413176.1
-
15
1487
DQ413177.1
-
11
1200
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
actin cytoskeleton organization
actin binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0