Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GON7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032490.4 | NP_115879.2 | 1 | 221 | ||
GenBank mRNA | BC021733.2 | - | 1 | 201 | 4796574 | |
BC065216.1 | - | 1 | 123 | |||
BC107429.1 | - | 1 | 189 | 4809642 | ||
BC109390.1 | - | 1 | 132 | |||
BF131439.1 | - | 1 | 221 |