Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039374.4 | NP_001034463.4 | 4 | 378 | ||
GenBank mRNA | AK092639.1 | - | 1 | 636 | ||
BC064582.1 | - | 4 | 366 | |||
BC137317.1 | - | 4 | 353 | 9021314 | ||
BC137318.1 | - | 4 | 353 | 9021315 | ||
BF979345.1 | - | 293 | ||||
DB446839.1 | - | 4 | 378 |