Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003705.4 | NP_003696.2 | 8 | 891 | ||
NR_047549.1 | 7 | 775 | ||||
XM_011512070.2 | XP_011510372.1 | 9 | 963 | |||
GenBank mRNA | AJ496568.1 | - | 8 | 820 | ||
AK022828.1 | - | 8 | 829 | |||
AK091071.1 | - | 7 | 692 | |||
BC016932.1 | - | 8 | 808 | 4420100 | ||
Y14494.1 | - | 8 | 822 |