Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003713.4 | NP_003704.3 | 5, 6 | 1368 | ||
GenBank mRNA | AB000889.1 | - | 5, 6 | 806 | ||
AF017786.1 | - | 6, 7 | 1174 | |||
AF043329.1 | - | 5, 6 | 1093 | |||
AF480883.1 | - | 6, 7 | 1180 | |||
AK123105.1 | - | 1, 2 | 1786 | |||
AK312439.1 | - | 5, 6 | 1326 | |||
BC009196.2 | - | 5, 6 | 1171 | 3960223 | ||
BT019589.1 | - | 5, 6 | 800 | |||
U79294.1 | - | 6, 7 | 1173 |