Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037283.1 | NP_001032360.1 | 18 | 2479 | ||
NM_003751.3 | NP_003742.2 | 18 | 2479 | |||
XM_011515599.1 | XP_011513901.1 | 18 | 2778 | |||
XM_011515600.1 | XP_011513902.1 | 18 | 2778 | |||
XM_011515601.1 | XP_011513903.1 | 19 | 2661 | |||
XM_017012752.1 | XP_016868241.1 | 18 | 2778 | |||
GenBank mRNA | AI888128.1 | - | 2 | 529 | ||
AK093807.1 | - | 4 | 1343 | |||
AK126670.1 | - | 4 | 3145 | |||
AK127885.1 | - | 3 | 1477 | |||
AK300967.1 | - | 19 | 2221 | |||
AK302058.1 | - | 19 | 2005 | |||
BC001173.1 | - | 18 | 2427 | 3501759 | ||
BC009986.1 | - | 17 | 1821 | |||
BC110865.1 | - | 18 | 2405 | 6175284 | ||
BQ015102.1 | - | 2 | 540 | |||
U62583.1 | - | 18 | 2449 | |||
U78525.1 | - | 18 | 2471 |