Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198622.1 | NP_001185551.1 | 1 | 893 | ||
NM_001198623.1 | NP_001185552.1 | 1 | 893 | |||
NM_001198624.1 | NP_001185553.1 | 2 | 201 | |||
NM_003808.3 | NP_003799.1 | 1 | 893 | |||
NM_172087.2 | NP_742084.1 | 1 | 893 | |||
NM_172088.2 | NP_742085.1 | 1 | 893 | |||
NR_073490.2 | 1 | 893 | ||||
GenBank mRNA | AF046888.1 | - | 1 | 426 | ||
AF114011.1 | - | 1 | 426 | |||
AF114012.1 | - | 1 | 342 | |||
AF114013.1 | - | 1 | 398 | |||
AF136294.1 | - | 1 | 145 | |||
AF184972.1 | - | 1 | 270 | |||
AK090698.1 | - | 1 | 709 | |||
AK225556.1 | - | 1 | 437 | |||
AK301221.1 | - | 1 | 426 | |||
AK314518.1 | - | 1 | 433 | |||
AY081050.1 | - | 1 | 417 | |||
AY358880.1 | - | 1 | 258 | |||
BC008042.2 | - | 1 | 248 | 3837834 | ||
BF793371.1 | - | 199 | ||||
BT019561.1 | - | 1 | 145 | |||
BT019562.1 | - | 1 | 145 | |||
CR542040.1 | - | 1 | 145 | |||
CR542073.1 | - | 1 | 145 | |||
DQ149579.1 | - | 1 | 175 | |||
EF211088.1 | - | 1 | 175 |