Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006773.3 | NP_006764.3 | 8, 9 | 1315 | ||
GenBank mRNA | AB209392.1 | - | 1, 2 | 382 | ||
AK001467.1 | - | 8, 9 | 1287 | |||
AK091227.1 | - | 6, 7 | 742 | |||
BC001238.1 | - | 8, 9 | 1244 | 3452935 | ||
BC003360.1 | - | 8, 9 | 1274 | 2900252 | ||
BC024739.1 | - | 8, 9 | 1289 | 4995871 | ||
CR457060.1 | - | 7, 8 | 1036 | |||
X98743.1 | - | 7, 8 | 1053 |