Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005407.1 | NP_001005407.1 | 3 | 563 | ||
NM_021098.2 | NP_066921.2 | 3 | 563 | |||
XM_005255652.4 | XP_005255709.1 | 3 | 409 | |||
XM_006720963.3 | XP_006721026.1 | 3 | 409 | |||
XM_006720964.3 | XP_006721027.1 | 3 | 409 | |||
XM_006720965.3 | XP_006721028.1 | 3 | 409 | |||
XM_006720967.3 | XP_006721030.1 | 3 | 410 | |||
XM_006720968.3 | XP_006721031.1 | 3 | 410 | |||
XM_011522727.2 | XP_011521029.1 | 3 | 416 | |||
XM_017023819.1 | XP_016879308.1 | 3 | 409 | |||
XM_017023820.1 | XP_016879309.1 | 3 | 409 | |||
XM_017023821.1 | XP_016879310.1 | 3 | 409 | |||
GenBank mRNA | AF051946.3 | - | 3 | 394 | ||
AF073931.1 | - | 3 | 563 | |||
AJ420779.1 | - | 2 | 315 |