Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271602.1 | NP_001258531.1 | 10, 11 | 1253 | ||
NM_014606.2 | NP_055421.1 | 11, 12 | 1508 | |||
XM_005263327.3 | XP_005263384.1 | 12, 13 | 1635 | |||
XM_005263329.3 | XP_005263386.1 | 11, 12 | 1508 | |||
XM_005263330.3 | XP_005263387.1 | 11, 12 | 1508 | |||
XM_005263331.1 | XP_005263388.1 | 11, 12 | 1508 | |||
XM_005263332.3 | XP_005263389.1 | 11, 12 | 1508 | |||
XM_017008806.1 | XP_016864295.1 | 12, 13 | 1635 | |||
XM_017008807.1 | XP_016864296.1 | 10, 11 | 1231 | |||
XM_017008808.1 | XP_016864297.1 | 3, 4 | 314 | |||
GenBank mRNA | AK289990.1 | - | 11, 12 | 1451 | ||
AK296379.1 | - | 10, 11 | 1229 | |||
D25215.1 | - | 11, 12 | 1435 |