Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286506.1 | NP_001273435.1 | 3 | 403 | ||
NM_080860.3 | NP_543136.1 | 4 | 517 | |||
XM_005261208.2 | XP_005261265.1 | 2 | 157 | |||
XM_011529786.1 | XP_011528088.1 | 4 | 409 | |||
GenBank mRNA | AB006536.1 | - | 4 | 517 | ||
AB041016.1 | - | 4 | 454 | |||
AK057315.1 | - | 4 | 410 | |||
AK314746.1 | - | 4 | 354 | |||
BC101519.1 | - | 4 | 396 | 8069025 | ||
BC113367.1 | - | 4 | 396 | 8322419 | ||
BG822861.1 | - | 238 |