Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS4 SIRNA Assays ›
Gene Symbol:
HPS4
Gene Name:
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases:
BLOC3S2, LE
Chromosome Location:
Chr.22: 26443613 - 26483863 on Build GRCh38
UniGene ID:
Hs.474436
Species:
Human
siRNA ID
HSS189688
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022081.5
NP_071364.4
11
2076
NM_152841.2
NP_690054.1
9
1693
NR_073135.1
11
2094
NR_073136.1
10
1856
XM_006724353.3
XP_006724416.1
12
2121
XM_006724354.3
XP_006724417.1
12
2026
XM_006724360.3
XP_006724423.1
7
1304
XM_011530485.1
XP_011528787.1
13
2199
XM_011530486.2
XP_011528788.1
13
2104
XM_011530487.2
XP_011528789.1
14
2309
XM_011530488.2
XP_011528790.1
13
2107
XM_011530489.2
XP_011528791.1
14
2210
XM_011530490.2
XP_011528792.1
12
2145
XM_011530491.2
XP_011528793.1
13
2199
XM_011530492.1
XP_011528794.1
13
2199
XM_011530493.2
XP_011528795.1
13
2199
XM_011530494.2
XP_011528796.1
11
1495
XM_011530495.2
XP_011528797.1
5
1001
XM_011530496.2
XP_011528798.1
6
836
XM_017029042.1
XP_016884531.1
11
1972
XM_017029043.1
XP_016884532.1
12
2177
XM_017029044.1
XP_016884533.1
11
1975
XM_017029045.1
XP_016884534.1
12
2145
XM_017029046.1
XP_016884535.1
11
2067
XM_017029047.1
XP_016884536.1
12
2145
XM_017029048.1
XP_016884537.1
11
2067
XM_017029049.1
XP_016884538.1
11
1972
XM_017029050.1
XP_016884539.1
11
2067
XM_017029051.1
XP_016884540.1
11
1972
XM_017029052.1
XP_016884541.1
12
2230
XM_017029053.1
XP_016884542.1
4
1589
XM_017029054.1
XP_016884543.1
11
2176
XM_017029055.1
XP_016884544.1
11
2030
XM_017029056.1
XP_016884545.1
12
2243
XM_017029057.1
XP_016884546.1
12
2092
XM_017029058.1
XP_016884547.1
12
2126
XM_017029059.1
XP_016884548.1
12
2001
XM_017029060.1
XP_016884549.1
12
2000
XM_017029061.1
XP_016884550.1
12
2201
XM_017029062.1
XP_016884551.1
10
1976
XM_017029063.1
XP_016884552.1
12
2110
XM_017029064.1
XP_016884553.1
10
1859
GenBank mRNA
AB051454.1
-
10
4266
AK054717.1
-
8
2174
AK095406.1
-
1
613
AK123691.1
-
6
1803
AK290211.1
-
11
1996
AL365512.1
-
9
1597
AL713795.1
-
11
2025
AY043416.1
-
10
1596
BC028625.1
-
11
2090
BC050285.1
-
11
2067
BC065030.1
-
10
1798
6148557
CR456392.1
-
10
1497
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein targeting
lysosome organization
blood coagulation
hemostasis
melanocyte differentiation
positive regulation of eye pigmentation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein binding
protein homodimerization activity
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0