Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NAV2 SIRNA Assays ›
Gene Symbol:
NAV2
Gene Name:
neuron navigator 2
Gene Aliases:
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
Chromosome Location:
Chr.11: 19345200 - 20121601 on Build GRCh38
UniGene ID:
Hs.502116
Species:
Human
siRNA ID
HSS189694
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NAV2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001111018.1
NP_001104488.1
26, 27
5541
NM_001111019.2
NP_001104489.1
16, 17
2953
NM_001244963.1
NP_001231892.1
29, 30
6042
NM_145117.4
NP_660093.2
26, 27
5865
NM_182964.5
NP_892009.3
27, 28
5874
XM_005253214.4
XP_005253271.1
15, 16
7349
XM_006718364.3
XP_006718427.1
28, 29
6295
XM_006718365.3
XP_006718428.1
28, 29
6265
XM_006718366.3
XP_006718429.1
29, 30
5528
XM_006718367.3
XP_006718430.1
18, 19
7562
XM_006718368.3
XP_006718431.1
15, 16
7385
XM_006718369.2
XP_006718432.1
18, 19
3057
XM_011520444.2
XP_011518746.1
28, 29
6355
XM_011520445.2
XP_011518747.1
28, 29
6298
XM_011520446.2
XP_011518748.1
27, 28
6256
XM_011520447.2
XP_011518749.1
27, 28
6253
XM_011520448.2
XP_011518750.1
29, 30
6235
XM_011520449.2
XP_011518751.1
26, 27
6190
XM_011520450.2
XP_011518752.1
26, 27
6154
XM_011520451.2
XP_011518753.1
28, 29
5742
XM_011520452.2
XP_011518754.1
27, 28
6004
XM_011520453.2
XP_011518755.1
18, 19
2992
XM_011520454.2
XP_011518756.1
15, 16
2782
XM_017018520.1
XP_016874009.1
30, 31
6156
XM_017018521.1
XP_016874010.1
25, 26
6145
XM_017018522.1
XP_016874011.1
29, 30
6112
XM_017018523.1
XP_016874012.1
29, 30
7041
XM_017018524.1
XP_016874013.1
29, 30
5864
XM_017018525.1
XP_016874014.1
23, 24
4707
XM_017018526.1
XP_016874015.1
19, 20
5343
GenBank mRNA
AB037840.3
-
27, 28
5875
AB063115.1
-
26, 27
5541
AB063116.1
-
26, 27
5834
AF466143.1
-
26, 27
5580
AF466144.1
-
27, 28
5589
AK074287.1
-
16, 17
2953
BC016054.1
-
8, 9
858
BC150290.1
-
27, 28
5875
8860130
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure by baroreceptor feedback
sensory perception of sound
sensory perception of smell
locomotory behavior
optic nerve development
glossopharyngeal nerve development
vagus nerve development
helicase activity
protein binding
ATP binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0