Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG16L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318766.1 | NP_001305695.1 | 14 | 1540 | ||
NM_033388.1 | NP_203746.1 | 14 | 1420 | |||
XM_005274376.4 | XP_005274433.1 | 14 | 1419 | |||
XM_006718732.2 | XP_006718795.1 | 13 | 1356 | |||
XM_006718733.3 | XP_006718796.1 | 11 | 2123 | |||
XM_006718734.2 | XP_006718797.1 | 6 | 2599 | |||
XM_011545332.1 | XP_011543634.1 | 13 | 1305 | |||
XM_011545333.1 | XP_011543635.1 | 14 | 1868 | |||
XM_011545334.1 | XP_011543636.1 | 14 | 1281 | |||
GenBank mRNA | AK024423.1 | - | 5 | 3634 | ||
AK090597.1 | - | 14 | 1531 | |||
AK091151.1 | - | 13 | 3008 | |||
AK093017.1 | - | 14 | 1393 | |||
AL832974.1 | - | 6 | 1031 | |||
BC036713.2 | - | 12 | 1149 | |||
BC137489.1 | - | 14 | 1518 | 9021489 | ||
BC137490.1 | - | 14 | 1518 | 9021490 | ||
BC142718.1 | - | 12 | 3116 | |||
BC146660.1 | - | 12 | 3116 |