Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305163.1 | NP_001292092.1 | 11 | 2131 | ||
NM_001305164.1 | NP_001292093.1 | 11 | 1510 | |||
NM_133368.2 | NP_588609.1 | 11 | 1532 | |||
XM_005256220.1 | XP_005256277.1 | 11 | 1839 | |||
XM_011523427.1 | XP_011521729.1 | 12 | 1618 | |||
XM_011523428.1 | XP_011521730.1 | 12 | 1932 | |||
XM_011523430.1 | XP_011521732.1 | 12 | 1559 | |||
XM_017023844.1 | XP_016879333.1 | 12 | 2305 | |||
GenBank mRNA | AB075852.1 | - | 11 | 1452 | ||
AK027549.1 | - | 11 | 2082 | |||
AK124601.1 | - | 3 | 2449 | |||
AK172845.1 | - | 11 | 1488 | |||
BC013173.2 | - | 11 | 1487 | 4340287 |