Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168299.1 | NP_001161771.1 | 6 | 1293 | ||
NM_001168300.1 | NP_001161772.1 | 5 | 1541 | |||
NM_001168301.1 | NP_001161773.1 | 6 | 1191 | |||
NM_001168302.1 | NP_001161774.1 | 6 | 1485 | |||
NM_001168303.1 | NP_001161775.1 | 5 | 1411 | |||
NM_033495.3 | NP_277030.2 | 5 | 2061 | |||
XM_011531409.2 | XP_011529711.1 | 7 | 1953 | |||
XM_011531410.2 | XP_011529712.1 | 7 | 1253 | |||
XM_011531411.1 | XP_011529713.1 | 6 | 1632 | |||
XM_017029950.1 | XP_016885439.1 | 5 | 2322 | |||
GenBank mRNA | AB037730.1 | - | 4 | 3420 | ||
AK122724.1 | - | 5 | 1128 | |||
AK125356.1 | - | 5 | 2061 | |||
AK296324.1 | - | 6 | 1200 | |||
AK297523.1 | - | 5 | 1138 | |||
AK299257.1 | - | 5 | 1305 | |||
AK316509.1 | - | 6 | 1445 | |||
AL591986.1 | - | 5 | 1208 | |||
BC064576.1 | - | 5 | 1213 | 5727768 | ||
BP380009.1 | - | 1 | 35 |