Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034996.2 | NP_001030168.1 | 6 | 499 | ||
NM_003973.4 | NP_003964.3 | 6 | 435 | |||
GenBank mRNA | AK222750.1 | - | 1 | 394 | ||
AK223111.1 | - | 6 | 388 | |||
AK290674.1 | - | 6 | 395 | |||
BC000606.2 | - | 6 | 401 | 3346054 | ||
BC005134.2 | - | 5 | 366 | 3928410 | ||
BC009294.2 | - | 6 | 398 | 4123062 | ||
BC019651.1 | - | 1 | 373 | 4454086 | ||
BC022805.1 | - | 1 | 363 | |||
BC029036.1 | - | 6 | 487 | 5183728 | ||
BC071913.1 | - | 1 | 395 | 6671085 | ||
BG564298.1 | - | 6 | 435 | |||
BG776586.1 | - | 6 | 418 | |||
CD298951.1 | - | 6 | 502 | |||
D87735.1 | - | 6 | 383 | |||
DQ118667.1 | - | 5 | 366 | |||
L10376.1 | - | 1 | 371 | |||
U16738.1 | - | 5 | 372 | |||
U92982.1 | - | 5 | 300 |