Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKMYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258450.1 | NP_001245379.1 | 3 | 662 | ||
NM_001258451.1 | NP_001245380.1 | 3 | 733 | |||
NM_004203.4 | NP_004194.3 | 3 | 869 | |||
NM_182687.2 | NP_872629.1 | 3 | 869 | |||
XM_011522734.2 | XP_011521036.1 | 3 | 830 | |||
XM_011522735.2 | XP_011521037.1 | 3 | 656 | |||
XM_011522736.2 | XP_011521038.1 | 2 | 663 | |||
GenBank mRNA | AK097642.1 | - | 3 | 618 | ||
AK098452.1 | - | 3 | 721 | |||
AK294379.1 | - | 2 | 630 | |||
AK301926.1 | - | 3 | 538 | |||
AK303005.1 | - | 3 | 747 | |||
AK309964.1 | - | 2 | 663 | |||
BC121161.1 | - | 2 | 557 | |||
BC121162.1 | - | 2 | 557 | |||
BC130011.1 | - | 2 | 557 | |||
BC153823.1 | - | 2 | 557 | |||
U56816.1 | - | 2 | 573 |