Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304284.1 | NP_001291213.1 | 10 | 2275 | ||
NM_004505.3 | NP_004496.2 | 2 | 1744 | |||
XM_011524050.1 | XP_011522352.1 | 6 | 2493 | |||
XM_011524051.2 | XP_011522353.1 | 8 | 1588 | |||
XM_011524052.2 | XP_011522354.1 | 7 | 1702 | |||
XM_011524053.2 | XP_011522355.1 | 8 | 1615 | |||
XM_011524054.2 | XP_011522356.1 | 8 | 1582 | |||
XM_011524055.2 | XP_011522357.1 | 6 | 1509 | |||
XM_011524056.2 | XP_011522358.1 | 3 | 1462 | |||
XM_011524057.2 | XP_011522359.1 | 7 | 1691 | |||
XM_011524058.2 | XP_011522360.1 | 7 | 1691 | |||
XM_011524059.2 | XP_011522361.1 | 7 | 1691 | |||
XM_017025290.1 | XP_016880779.1 | 5 | 2331 | |||
GenBank mRNA | AB449915.1 | - | 1 | 45 | ||
AY143550.1 | - | 2 | 1744 | |||
BX647719.1 | - | 10 | 2276 | |||
X63546.1 | - | 2 | 1741 | |||
X63547.1 | - | 2 | 1741 |