Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHURC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204063.1 | NP_001190992.1 | 2 | 200 | ||
NM_001204064.1 | NP_001190993.1 | 2 | 200 | |||
NM_145165.3 | NP_660148.3 | 2 | 200 | |||
GenBank mRNA | AF060510.1 | - | 2 | 118 | ||
AK057626.1 | - | 2 | 111 | |||
AK296141.1 | - | 2 | 84 | |||
AK315905.1 | - | 2 | 118 | |||
AV734503.1 | - | 2 | 200 | |||
BC020550.1 | - | 2 | 149 | 4155545 | ||
BP305870.1 | - | 2 | 130 |
Gene Symbol: CHURC1-FNTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202558.1 | NP_001189487.1 | 2 | 200 | ||
NM_001202559.1 | NP_001189488.1 | 2 | 200 | |||
GenBank mRNA | AK296850.1 | - | 2 | 68 | ||
AK303739.1 | - | 2 | 76 |