Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282382.1 | NP_001269311.1 | 5 | 994 | ||
NM_001282383.1 | NP_001269312.1 | 6 | 848 | |||
NM_004219.3 | NP_004210.1 | 6 | 687 | |||
GenBank mRNA | AF075242.1 | - | 5 | 611 | ||
AF095287.1 | - | 6 | 627 | |||
AJ223953.1 | - | 5 | 696 | |||
AK312209.1 | - | 6 | 665 | |||
BC101834.1 | - | 6 | 620 | 8069340 | ||
BC112071.1 | - | 6 | 620 | 8327539 | ||
BC128193.1 | - | 5 | 591 | 40113885 | ||
BC128194.1 | - | 5 | 591 | 40113886 | ||
BM990391.1 | - | 1 | 96 | |||
BU182149.1 | - | 6 | 844 | |||
CR457135.1 | - | 5 | 591 | |||
CR541685.1 | - | 5 | 591 |
Gene Symbol: PTTG3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF095289.1 | - | 1 | 591 |