Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 8 | 518 | ||
NM_001012632.1 | NP_001012650.1 | 7 | 317 | |||
NM_001012633.1 | NP_001012651.1 | 7 | 451 | |||
NM_001012634.1 | NP_001012652.1 | 6 | 258 | |||
NM_001012635.1 | NP_001012653.1 | 6 | 253 | |||
NM_001012636.1 | NP_001012654.1 | 5 | 241 | |||
NM_001012718.1 | NP_001012736.1 | 7 | 454 | |||
NM_001308078.1 | NP_001295007.1 | 6 | 452 | |||
NM_004221.4 | NP_004212.4 | 7 | 451 | |||
GenBank mRNA | AK296143.1 | - | 6 | 263 | ||
AY495331.1 | - | 7 | 441 | |||
AY495332.1 | - | 7 | 441 | |||
AY495333.1 | - | 6 | 399 | |||
BC009401.2 | - | 6 | 589 | 4300407 | ||
BC018782.1 | - | 7 | 298 | 4873608 | ||
BC105602.1 | - | 7 | 319 | 6496101 | ||
BF568456.1 | - | 6 | 590 | |||
BG748458.1 | - | 7 | 397 | |||
BM759359.1 | - | 4 | 192 | |||
BM822755.1 | - | 8 | 426 | |||
EU339107.1 | - | 4 | 120 | |||
FJ985780.1 | - | 6 | 345 | |||
JN546102.1 | - | 5 | 180 | |||
M59807.1 | - | 6 | 437 |