Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FANK1 SIRNA Assays ›
Gene Symbol:
FANK1
Gene Name:
fibronectin type III and ankyrin repeat domains 1
Gene Aliases:
HSD13
Chromosome Location:
Chr.10: 125896539 - 126009599 on Build GRCh38
UniGene ID:
Hs.352591
Species:
Human
siRNA ID
HSS190114
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_145235.3
NP_660278.3
10, 11
1081
XM_005270280.3
XP_005270337.1
9, 10
897
XM_006718058.3
XP_006718121.1
12, 13
1080
XM_006718064.3
XP_006718127.1
11, 12
1074
XM_011540348.2
XP_011538650.1
12, 13
1195
XM_011540349.2
XP_011538651.1
12, 13
1199
XM_011540350.2
XP_011538652.1
11, 12
1158
XM_011540351.2
XP_011538653.1
11, 12
1134
XM_011540356.2
XP_011538658.1
10, 11
1046
XM_011540360.2
XP_011538662.1
10, 11
975
XM_011540361.2
XP_011538663.1
10, 11
910
XM_011540362.2
XP_011538664.1
8, 9
796
XM_017016916.1
XP_016872405.1
12, 13
1141
XM_017016917.1
XP_016872406.1
12, 13
1276
XM_017016918.1
XP_016872407.1
11, 12
1083
XM_017016919.1
XP_016872408.1
11, 12
1235
XM_017016920.1
XP_016872409.1
12, 13
1292
XM_017016921.1
XP_016872410.1
11, 12
1198
XM_017016922.1
XP_016872411.1
11, 12
1098
XM_017016924.1
XP_016872413.1
13, 14
1253
XM_017016926.1
XP_016872415.1
7, 8
1210
GenBank mRNA
AY358154.1
-
11, 12
1137
BC024189.2
-
10, 11
1081
4826296
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0