Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287489.1 | NP_001274418.1 | 30, 31 | 3986 | ||
NM_004802.3 | NP_004793.2 | 13, 14 | 1784 | |||
NM_194248.2 | NP_919224.1 | 30, 31 | 3986 | |||
NM_194322.2 | NP_919303.1 | 12, 13 | 1938 | |||
NM_194323.2 | NP_919304.1 | 13, 14 | 1784 | |||
XM_017005338.1 | XP_016860827.1 | 30, 31 | 3799 | |||
GenBank mRNA | AF107403.1 | - | 13, 14 | 1784 | ||
AF183185.1 | - | 30, 31 | 3986 | |||
AF183186.1 | - | 12, 13 | 1938 | |||
AF183187.1 | - | 13, 14 | 1784 | |||
AK296272.1 | - | 3, 4 | 341 |