Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052957.4 | NP_443189.1 | 12 | 2344 | ||
XM_006724716.3 | XP_006724779.1 | 12 | 1897 | |||
XM_011531063.2 | XP_011529365.1 | 11 | 2164 | |||
XM_011531064.1 | XP_011529366.1 | 12 | 1947 | |||
XM_011531066.2 | XP_011529368.1 | 9 | 2083 | |||
XM_011531067.2 | XP_011529369.1 | 9 | 2083 | |||
XM_017029965.1 | XP_016885454.1 | 13 | 1998 | |||
GenBank mRNA | AJ311392.1 | - | 11 | 1845 | ||
AK097433.1 | - | 12 | 1958 | |||
AK127607.1 | - | 11 | 2380 | |||
BC136259.1 | - | 12 | 1912 | 9020246 |