Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172766.2 | NP_001166237.1 | 15 | 2181 | ||
NM_014491.3 | NP_055306.1 | 15 | 2184 | |||
NM_148898.3 | NP_683696.2 | 16 | 2259 | |||
NM_148900.3 | NP_683698.2 | 16 | 2235 | |||
NR_033766.1 | 18 | 2195 | ||||
NR_033767.1 | 16 | 2242 | ||||
XM_017012801.1 | XP_016868290.1 | 18 | 2865 | |||
GenBank mRNA | AF337817.1 | - | 15 | 1960 | ||
AK131266.1 | - | 18 | 2179 | |||
AK296957.1 | - | 16 | 2115 | |||
AY144615.1 | - | 16 | 2254 | |||
BC126104.1 | - | 16 | 1916 | 8991820 | ||
BC143866.1 | - | 16 | 1916 | 9052382 | ||
BC143867.1 | - | 15 | 1855 | 9052383 | ||
CR749236.1 | - | 22 | 2565 | |||
DQ778626.1 | - | 10 | 1255 | |||
KJ534839.1 | - | 15 | 1885 | |||
KJ535079.1 | - | 14 | 2023 | |||
KJ535086.1 | - | 14 | 1809 |