Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004260.3 | NP_004251.3 | 5 | 601 | ||
XM_011517384.2 | XP_011515686.1 | 5 | 601 | |||
XM_017013991.1 | XP_016869480.1 | 5 | 606 | |||
XM_017013992.1 | XP_016869481.1 | 5 | 605 | |||
XM_017013993.1 | XP_016869482.1 | 5 | 605 | |||
XM_017013994.1 | XP_016869483.1 | 5 | 607 | |||
XM_017013995.1 | XP_016869484.1 | 5 | 603 | |||
XM_017013996.1 | XP_016869485.1 | 5 | 604 | |||
XM_017013997.1 | XP_016869486.1 | 5 | 606 | |||
XM_017013998.1 | XP_016869487.1 | 5 | 603 | |||
XM_017013999.1 | XP_016869488.1 | 5 | 605 | |||
XM_017014001.1 | XP_016869490.1 | 3 | 701 | |||
GenBank mRNA | AB006532.1 | - | 5 | 559 | ||
CN286060.1 | - | 5 | 593 | |||
KJ147518.1 | - | 1 | 106 |