Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004814.2 | NP_004805.2 | 6 | 739 | ||
GenBank mRNA | AF090988.1 | - | 6 | 708 | ||
AK074516.1 | - | 6 | 722 | |||
AK123103.1 | - | 2 | 1149 | |||
AK298823.1 | - | 6 | 757 | |||
AK313800.1 | - | 6 | 704 | |||
BC001494.2 | - | 6 | 715 | 3504573 | ||
EF011621.1 | - | 6 | 714 |