Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPEPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006310.3 | NP_006301.3 | 16 | 2012 | ||
XM_011525496.1 | XP_011523798.1 | 17 | 1814 | |||
XM_017025372.1 | XP_016880861.1 | 16 | 1974 | |||
XM_017025373.1 | XP_016880862.1 | 17 | 1814 | |||
GenBank mRNA | AJ132583.1 | - | 16 | 1742 | ||
AK095877.1 | - | 17 | 1778 | |||
AK096491.1 | - | 1 | 470 | |||
AK096709.1 | - | 18 | 1799 | |||
AK293450.1 | - | 10 | 944 | |||
AK293995.1 | - | 9 | 926 | |||
AK296887.1 | - | 15 | 1655 | |||
AK297107.1 | - | 15 | 1516 | |||
AK303037.1 | - | 17 | 1807 | |||
BC065294.1 | - | 16 | 1701 | 6059589 |