Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARTPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004291.3 | NP_004282.1 | 1, 2 | 281 | ||
GenBank mRNA | BC029882.1 | - | 1, 2 | 277 | 5171665 | |
CR542216.1 | - | 1, 2 | 150 | |||
U16826.1 | - | 1, 2 | 169 |