Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304447.1 | NP_001291376.1 | 3 | 1079 | ||
NM_014653.3 | NP_055468.2 | 2 | 1248 | |||
XM_017020243.1 | XP_016875732.1 | 3 | 1533 | |||
XM_017020244.1 | XP_016875733.1 | 4 | 1694 | |||
XM_017020245.1 | XP_016875734.1 | 4 | 1191 | |||
XM_017020246.1 | XP_016875735.1 | 2 | 922 | |||
XM_017020247.1 | XP_016875736.1 | 3 | 1065 | |||
XM_017020248.1 | XP_016875737.1 | 3 | 1537 | |||
XM_017020249.1 | XP_016875738.1 | 4 | 1694 | |||
XM_017020250.1 | XP_016875739.1 | 2 | 1248 | |||
XM_017020251.1 | XP_016875740.1 | 3 | 1059 | |||
XM_017020252.1 | XP_016875741.1 | 3 | 1537 | |||
GenBank mRNA | AB018332.2 | - | 2 | 985 | ||
AK293764.1 | - | 1 | 507 | |||
BC110330.1 | - | 2 | 1031 | 5735001 | ||
BC136687.1 | - | 1 | 777 | 9020677 | ||
BC136688.1 | - | 1 | 777 | 9020678 | ||
BX647283.1 | - | 3 | 1030 |