Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014671.2 | NP_055486.2 | 20 | 3177 | ||
XM_005249564.4 | XP_005249621.1 | 19 | 3111 | |||
XM_017012818.1 | XP_016868307.1 | 21 | 3449 | |||
GenBank mRNA | AK127280.1 | - | 5 | 3448 | ||
AK295138.1 | - | 20 | 2857 | |||
D13635.1 | - | 20 | 3168 |